Dura Flock® Flock-Lined Nitrile Gloves, Microflex®, Ansell
Supplier: Microflex
CE Compliant
Certificates
About this item
Ambidextrous, powder-free, 100% nitrile gloves feature an ultra-thin absorbent flock lining to keep hands dry, comfortable, and protected.
- Beaded cuffs
- Fully textured for enhanced grip
- Comfortable grip for reduced hand fatigue
8-millimeter gloves are thicker than standard disposable gloves for increased durability and superior protection.
Specifications
- Catalog No:
- CA89094-568
- CA89094-570
- CA89094-574
- CA89094-572
- CA89094-576
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- Finger thickness:
- 8 mil
- 8 mil
- 8 mil
- 8 mil
- 8 mil
- Palm thickness:
- 8 mil
- 8 mil
- 8 mil
- 8 mil
- 8 mil