Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%) ACS, meets analytical specification of USP for HPLC, for spectrophotometry
Supplier: Burdick & Jackson
Certificates
About this item
Synonyms: Hexanes
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MW:86.18 g/mol
Boiling Pt:7.7 %(V)
Melting Pt:-95 °C
Density:0.659-0.673 g/cm³ (20 °C)
Flash Pt:-26 °C
MDL Number:MFCD02179311
ADR:3,II
Merck Index:13,04712
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Boiling Pt:
:Boiling Pt: 7.7 %(V)
:Melting Pt:
:Melting Pt: -95 °C
:Flash Pt:
:Flash Pt: -26 °C
:M.W. 86.18 g/mol
Storage Temperature:Ambient
EINECS:203-777-6
:7.7 %(V)
UN:1208
:3
:II
:-26 °C
:GHS07
:GHS08
:GHS02
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Color (APHA):10 max.
- Water-Soluble Titrable Acid:0.3 µeq/g max.
- GC Analysis (n-Hexane):60 % min.
- GC Analysis (total C6 isomers):99.9 % min.
- Residue:3 mg/L max.
- Sulfur Compounds (as S):0.005 % max.
- Thiophene:Passes test
- UV Absorbance @ 225 nm:0.05 AU max.
- UV Cutoff:195 nm max.
- Water by Karl Fisher Titration:0.01 % max.
- Hexane (mixture of isomers) ≥99.9% (n-Hexane > 60%) ACS, meets analytical specification of USP for HPLC, for spectrophotometry:
Specification Test Results
Related Information
Certificates
Search by Lot NumberHB Honeywell B
Datasheet
VWR GHS Compliance Letter (US)
Additional Product Details
VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Customers Who Bought This Also Bought