Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
Formula:HOCH₂CH(OH)CH₂OH
MW:92.09 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³
Flash Pt:177 °C
MDL Number:MFCD00004722
CAS Number:56-81-5
Merck Index:13,04497
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C
:M.W. 92.09 g/mol
EINECS:200-289-5
:177 °C
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (C₃H₅(OH)₃) (by GC, corrected for water):≥99.5 %
- ACS - Acrolein and Glucose:Passes Test
- ACS - Chlorinated Compounds (as Cl):≤0.003 %
- ACS - Color (APHA):≤10
- ACS - Fatty Acid Esters (as Butyric Acid):≤0.05 %
- ACS - Heavy Metals (as Pb):≤2 ppm
- ACS - Neutrality:Passes Test
- ACS - Residue after Ignition:≤0.005 %
- Solubility (5 in 50):Passes Test
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Sulfate (SO₄):≤0.001 %
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- Appearance (clear, colorless, syrupy liquid):
- Macron Fine Chemicals™, Glycerine ≥99.5% (by GC, corrected for water content), AR® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number5092 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 5092.pdf
Datasheet
Customers Who Bought This Also Bought