Guanidinium hydrochloride ≥99.5%, NF-GenAR® for biotechnology, Macron Fine Chemicals™
Supplier: Avantor
Certificates
About this item
Synonyms: Guanadine hydrochloride,Guanidine chloride,Guanidinium chloride
Formula:NH₂C(=NH)NH₂·HCl
MW:95.53 g/mol
Melting Pt:185 °C
Density:1.354 g/cm³ (20 °C)
MDL Number:MFCD00013026
CAS Number:50-01-1
Merck Index:13,04578
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 185 °C
:Flash Pt:
:Flash Pt:
:M.W. 95.53 g/mol
:Guanidinium hydrochloride
EINECS:200-002-3
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Assay (NH₂C(:NH)NH₂·HCl):≥99.5 %
- Acidity (as HCl):≤0.01 %
- Arsenic and Antimony (as As):≤10 ppm
- Copper (Cu):≤10 ppm
- Heavy Metals (as Pb):≤10 ppm
- Iron (Fe):≤10 ppm
- Loss on Drying at 105°C:≤0.5 %
- Enzyme Activity - DNase Activity:Passes Test
- Enzyme Activity - RNase Activity:Passes Test
- Enzyme Activity - Protease Activity:Passes Test
- Melamine and Related Compounds:Passes Test
- Absorbance of a 6M Solution (260 nm) (au):≤0.030
- Residue after Ignition:≤0.20 %
- Water-Insoluble Matter:≤0.3 %
- Product Information (not a specification): Appearance - colorless:
- crystals:
- MANUFACTURED EXCLUSIVELY FOR AVANTOR PERFORMANCE MATERIALS, LLC., BY:
- BIOSPECTRA, INC., STROUDSBURG, PA:
- Macron Fine Chemicals™, Guanidinium hydrochloride ≥99.5%, NF-GenAR® for biotechnology:
Specification Test Results
Related Information
Certificates
Search by Lot Number0072 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 0072.pdf
Datasheet
sds_MACR0072
SDS
Customers Who Bought This Also Bought