Glycerine, anhydrous ≥99.5%, Ultrapure Bioreagent for biotechnology, for electrophoresis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
Formula:HOCH₂CH(OH)CH₂OH
MW:92.09 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
MDL Number:MFCD00004722
CAS Number:56-81-5
Merck Index:13,04497
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C
:M.W. 92.09 g/mol
EINECS:200-289-5
:177 °C
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (HOCH₂CHOHCH₂OH):≥99.5 %
- Absorbance of a 1M Solution - 280 nm:≤0.050
- DNase Activity (None Detected):Passes Test
- RNase Activity (None Detected):Passes Test
- Protease Activity (None Detected):Passes Test
- Absorbance of a 1M Solution - Color (APHA):≤10
- Residue after Ignition:≤0.005 %
- Fatty Acid Esters (as Butyric Acid):≤0.05 %
- Trace Impurities - Iron (Fe):≤5 ppm
- Trace Impurities - Lead (Pb):≤2.000 ppm
- Trace Impurities - Mercury (Hg):≤10.000 ppm
- Trace Impurities - Zinc (Zn):≤2.000 ppm
- Vegetable Based:
Specification Test Results
Related Information
Certificates
Search by Lot Number4043 Product Specifications
Additional Product Details
sds_BAKR4043
SDS
Customers Who Bought This Also Bought