Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
Formula:HOCH₂CH(OH)CH₂OH
MW:92.09 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
MDL Number:MFCD00004722
CAS Number:56-81-5
Merck Index:13,04497
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C
:M.W. 92.09 g/mol
EINECS:200-289-5
:177 °C
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets J.P. Chemical Specifications:
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B - Diethylene Glycol:≤0.10 %
- USP - Identification B - Ethylene Glycol:≤0.10 %
- USP - Identification C:Passes Test
- USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis):99.0 - 101.0 %
- USP - Specific Gravity at 25°/25°C:≥1.249
- USP - Color:Passes Test
- USP - Residue on Ignition:≤0.01 %
- USP - Chloride (Cl):≤0.001 %
- USP - Sulfate (SO₄):≤0.002 %
- USP - Chlorinated Compounds (as Cl):≤0.003 %
- USP - Fatty Acids and Esters:Passes Test
- USP - Water (H₂O):≤5.0 %
- USP - Related Compounds: Individual Impurity:≤0.1 %
- USP - Related Compo unds: Total Impurities:≤1.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Refractive Index, n²ºD:1.470 - 1.475
- EP/BP - Aldehydes:≤10 ppm
- EP/BP - Ester:Passes Test
- EP/BP - Halogenated Compounds:≤35 ppm
- EP/BP - Sugars:Passes Test
- EP/BP - Chloride (Cl):≤10 ppm
- EP/BP - Water (H₂O):≤2.0 %
- EP/BP - Ash (sulfated):≤0.01 %
- EP/BP - Impurity A and Related Substances - Impurity A:≤0.1 %
- EP/BP - Impurity A and Related Substances - Other Impurities with R^T<Glycerol:≤0.1 %
- EP/BP - Impurity A and Related Substances - Total Impurities R^T>Glycerol:≤0.5 %
- EP - Assay (anhydrous basis):98.0 - 101.0 %
- JP - Assay (anhydrous basis):98.0 - 101.0 %
- JP - Identification:Passes Test
- JP - Specific Gravity at 20°/20°C:≥1.258
- JP - Refractive Index, n²ºD:≥1.470
- JP - Color:Passes Test
- JP - Residue on Ignition:≤0.01 %
- JP - Chloride (Cl):≤0.001 %
- JP - Sulfate (SO₄):≤0.002 %
- JP - Heavy Metals (as Pb):≤5 ppm
- JP - Fatty Acids and Esters(as butyric acid):Passes Test
- JP - Readily Carbonizable Substances:Passes Test
- JP - Acidity or Alkalinity:Passes Test
- JP - Acrolein, Glucose, and Other Reducing Substances:Passes Test
- JP - Ammonium (NH₄):Passes Test
- JP - Calcium (Ca):Passes Test
- JP - Arsenic (As):≤2 ppm
- JP - Water (H₂O):≤2.0 %
- JP - Ethylene Glycol:≤0.1 %
- JP - Diethylene Glycol:≤0.1 %
- JP - Any Other Impurity:≤0.1 %
- JP - Total Impurities:≤1.0 %
- Endotoxin Concentration (EU/g):≤5
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number2143 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
sds_BAKR2143
SDS
spec_JT2143-1 spec_JT2143-3 spec_JT2143-7 spec_JT2143-9
Product specification
Customers Who Bought This Also Bought