Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
Formula:HOCH₂CH(OH)CH₂OH
MW:92.09 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
MDL Number:MFCD00004722
CAS Number:56-81-5
Merck Index:13,04497
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C
:M.W. 92.09 g/mol
EINECS:200-289-5
:177 °C
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (as HOCH₂CHOHCH₂OH) (calculated on anhydrous basis):99.0 - 101.0 %
- USP - Identification A:Passes Test
- USP - Identification B - Diethylene Glycol:≤0.10 %
- USP - Identification B - Ethylene Glycol:≤0.10 %
- USP - Identification C:Passes Test
- USP - Specific Gravity at 25°/25°C:≥1.249
- USP - Color:Passes Test
- USP - Residue on Ignition:≤0.01 %
- USP - Chloride (Cl):≤0.001 %
- USP - Sulfate (SO₄):≤0.002 %
- USP - Chlorinated Compounds (as Cl):≤0.003 %
- USP - Fatty Acids and Esters:Passes Test
- USP - Water (H₂O):≤5.0 %
- USP - Related Compounds: Individual Impurity:≤0.1 %
- USP - Related Compo unds: Total Impurities:≤1.0 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- J.T.Baker®, Glycerine (vegetable based) 99.0-101.0% (by anhydrous basis) USP:
Specification Test Results
Related Information
Certificates
Search by Lot Number2142 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
JTBaker 2142 Datasheet
Datasheet
spec_MK214201
Product specification
Customers Who Bought This Also Bought