Glycerine ≥99.5%, anhydrous ≥99.5% (by GC), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: 1,2,3-Propanetriol,Propane-1,2,3-triol
Formula:HOCH₂CH(OH)CH₂OH
MW:92.09 g/mol
Boiling Pt:290 °C (1013 hPa)
Melting Pt:18.6 °C
Density:1.26 g/cm³ (20 °C)
Flash Pt:177 °C
MDL Number:MFCD00004722
CAS Number:56-81-5
Merck Index:13,04497
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt: 290 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.6 °C
:Flash Pt:
:Flash Pt: 177 °C
:M.W. 92.09 g/mol
EINECS:200-289-5
:177 °C
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (HOCH₂CHOHCH₂OH) (by GC, corrected for water):≥99.5 %
- Color (APHA):≤10
- Residue after Ignition:≤0.005 %
- Neutrality:Passes Test
- Chlorinated Compounds (as Cl):≤0.003 %
- Sulfate (SO₄):≤0.001 %
- Acrolein and Glucose:Passes Test
- Fatty Acid Esters (as Butyric Acid):≤0.05 %
- Substances Darkened by H₂SO₄:Passes Test
- Water (H₂O)(by Karl Fischer titrn):≤0.5 %
- Heavy Metals (as Pb):≤2 ppm
- Meets Reagent Specifications for testing USP/NF monographs:
- Product Information (not specifications): Density (g/mL) at 25°C (typical) = 1.257:
- J.T.Baker®, Glycerine ≥99.5%, anhydrous ≥99.5% (by GC), BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number2136 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
Customers Who Bought This Also Bought