EDTA disodium salt 0.05 M (M/20) in aqueous solution, StandARd® volumetric solution, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Idranal III,Edetic acid disodium salt,Kestranal® 2S,Ethylenedinitrilotetra-acetic acid disodium salt,Disodium dihydrogen ethylenediaminetetraacetate
Formula:C₁₀H₁₄N₂Na₂O₈
MW:336.21 g/mol
MDL Number:MFCD00070672
CAS Number:139-33-3
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 336.21 g/mol
:EDTA disodium salt
Storage Temperature:Ambient
EINECS:205-358-3
:M.O.C. Category NI
:12,03556
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Molarity:0.0495 - 0.0505
- Macron Fine Chemicals™, EDTA disodium salt 0.05 M (M/20) in aqueous solution, StandARd® volumetric solution:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
H115 Product Specifications
Additional Product Details
sds_MACRH115
SDS
spec_MKH11505
Product specification
Customers Who Bought This Also Bought