Reagent alcohol 94-96%, AR® ACS (denatured with 5% isopropanol and 5% methanol), Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Reagent alcohol
MDL Number:MFCD00003568
CAS Number:64-17-5
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:94-96
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:Reagent alcohol
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
:3
:II
:GHS02
:GHS07
:GHS08
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Specially Denatured Alcohol Formula 3A (200 proof)(v/v):94.0 - 96.0 %
- Methanol (v/v):3.5 - 5.5 %
- Isopropyl Alcohol (2-propanol)(v/v):4.0 - 6.0 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- Specific Gravity at 25°C:0.782 - 0.793
- Water (H₂O):≤0.5 %
- SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200proof ethanol.:
- Appearance (clear, colorless liquid):
- Macron Fine Chemicals™, Reagent alcohol 94-96%, AR® ACS (denatured with 5% isopropanol and 5% methanol):
Specification Test Results
Related Information
Certificates
Search by Lot Number6183 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
sds_MACR6183
SDS
Customers Who Bought This Also Bought