EDTA disodium salt dihydrate 99.0-101.0%, AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate,Kestranal® 2S dihydrate,Disodium dihydrogen ethylenediaminetetraacetate dihydrate,Ethylenedinitrilotetra-acetic acid disodium salt dihydrate,Titriplex III,Ethylenediaminetetra-acetic acid disodium salt dihydrate,Kestranal® 2S 2 H2O,Edetate disodium dihydrate
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MW:372.24 g/mol
Melting Pt:255 °C
MDL Number:MFCD00150037
CAS Number:6381-92-6
Merck Index:13,03543
:EDTA disodium salt dihydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 255 °C
:Flash Pt:
:Flash Pt:
:M.W. 372.24 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (C₁₀H₁₄N₂Na₂O₈·2H₂O):99.0 - 101.0 %
- Identification A:Passes Test
- Identification B:Passes Test
- Identification C:Passes Test
- ACS - Heavy Metals (as Pb):≤0.005 %
- ACS - Insoluble Matter:≤0.005 %
- ACS - Iron (Fe):≤0.01 %
- ACS - Nitrilotriacetic Acid (by polarography):≤0.1 %
- ACS - pH of 5% Solution at 25°C:4.0 - 6.0
- Certificate Provided Reports Actual Lot Analysis:
- Macron Fine Chemicals™, EDTA disodium salt dihydrate 99.0-101.0%, AR® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number4931 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR4931
SDS
Customers Who Bought This Also Bought