EDTA disodium salt dihydrate 99.0-101.0% (by anhydrous basis) USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Ethylenediaminetetraacetic acid, disodium salt, dihydrate,Kestranal® 2S dihydrate,Disodium dihydrogen ethylenediaminetetraacetate dihydrate,Ethylenedinitrilotetra-acetic acid disodium salt dihydrate,Titriplex III,Ethylenediaminetetra-acetic acid disodium salt dihydrate,Kestranal® 2S 2 H2O,Edetate disodium dihydrate
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MW:372.24 g/mol
Melting Pt:255 °C
MDL Number:MFCD00150037
CAS Number:6381-92-6
Merck Index:13,03543
:EDTA disodium salt dihydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-101,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 255 °C
:Flash Pt:
:Flash Pt:
:M.W. 372.24 g/mol
Storage Temperature:Ambient
EINECS:205-358-3
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (C₁₀H₁₄N₂Na₂O₈) (anhydrous basis):99.0 - 101.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C:Passes Test
- USP - pH (1 in 20):4.0 - 6.0
- USP - Loss on Drying:8.7 - 11.4 %
- USP - Calcium (Ca):Passes Test
- USP - Limit of Nitrilotriacetic Acid:≤0.1 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Meets Reagent Specifications for testing USP/NF monographs:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number1395 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR1395
SDS
spec_MK139520
Product specification
Customers Who Bought This Also Bought