EDTA magnesium disodium salt hydrate, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Ethylenediaminetetra-acetic acid magensium disodium salt,Kestranal® 2S Mg,Magnesium disodium ethylenediaminetetraacetate,Ethylenedinitrilotetra-acetic acid magensium disodium salt,EDTA, Magnesium Derivative, Disodium Salt
CAS Number:14402-88-1
:MFCD00078217
Formula:C₁₀H₁₂MgN₂Na₂O₈·xH₂O
MW:358.5 g/mol (anhy)
MDL Number:MFCD03613601
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 358.5 g/mol (anhy)
:EDTA magnesium disodium salt hydrate
Storage Temperature:Ambient
EINECS:238-372-3
:GHS05
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Test for EDTA/Mg Ratio:Passes Test
Specification Test Results
Related Information
Customers Who Bought This Also Bought