Water, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Hydrogen oxide
Formula:H₂O
MW:18.02 g/mol
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
MDL Number:MFCD00011332
CAS Number:7732-18-5
Merck Index:12,10175
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 100 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
:Water
Storage Temperature:Ambient
EINECS:231-791-2
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.1 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤0.2 ppb
- pH:5.0 - 8.0
- Residue after Evaporation:≤0.0001 %
- Suitability for HPLC:Passes Test
- ACS - Specific Conductance at 25°C. (/uohms⁻¹ cm⁻¹):≤2.0
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤0.4 ppm
- ACS - Electrical Resistivity at 25°C (Mohm.cm):≥0.5
- Heavy Metals (as Pb):≤0.01 ppm
- Nitrate (NO₃):≤0.4 ppm
- Phosphate (PO₄):≤1.0 ppm
- Silicate (as SiO₂):≤0.01 ppm
- Sulfate (SO₄):≤1.0 ppm
- Filtered through a 0.2 micron filter.:
- Appearance (clear, colorless liquid):
- Macron Fine Chemicals™, Water, ChromAR® for liquid chromatography, for UV spectrophotometry:
Specification Test Results
Related Information
Certificates
Search by Lot Number6795 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 6795 MACR6795
Datasheet
sds_MACR6795
SDS
Customers Who Bought This Also Bought