Water, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Hydrogen oxide
Formula:H₂O
MW:18.02 g/mol
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
MDL Number:MFCD00011332
CAS Number:7732-18-5
Merck Index:12,10175
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 100 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
:Water
Storage Temperature:Ambient
EINECS:231-791-2
:0
:100
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Appearance:Passes Test
- Gradient Elution Test (a.u.) - 254 nm:≤0.002
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.1 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤0.2 ppb
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤50 ppb
- Trace Impurities - Aluminum (Al):≤50.0 ppb
- Trace Impurities - * Calcium (Ca):≤50.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Lithium (Li):≤30.0 ppb
- Trace Impurities - Magnesium (Mg):≤50.0 ppb
- Trace Impurities - Nickel (Ni):≤30.0 ppb
- Trace Impurities - Potassium (K):≤50 ppb
- Trace Impurities - * Sodium (Na):≤50.0 ppb
- * May change over time due to extraction from glass container.:
- Filtered through a 0.2 micron filter.:
- J.T.Baker®, Water, BAKER ANALYZED® for LC-MS:
Specification Test Results
Related Information
Certificates
Search by Lot Number9831 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals Avantor Top Rated Guide Production Insight Solutions for Controlled Environments and Manufacturing Processes Production insight v1 final1 VWR® Focus Production Vol 1 Issue 2
Product Literature
More
Brand Definition Baker Analyzed
Datasheet
Customers Who Bought This Also Bought