Ethyl acetate, BAKER ANALYZED® for LC-MS, J.T.Baker®
Catalog #: CAJT9828-3
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 141-78-6
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 l
- Cat. no.:CAJT9828-3
- Supplier No.:9828-03
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm:≤0.050
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm:≤1.00
- Assay (CH₃COOC₂H₅):≥99.6 %
- Residue after Evaporation:≤1.0 ppm
- Water (H₂O):≤0.05 %
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤50 ppb
- Trace Impurities - Aluminum (Al), For Information Only:
- Trace Impurities - * Calcium (Ca), For Information Only:
- Trace Impurities - Iron (Fe), For Information Only:
- Trace Impurities - Magnesium (Mg), For Information Only:
- Trace Impurities - Potassium (K), For Information Only:
- Trace Impurities - *Sodium (Na), For Information Only:
- * May change over time due to extraction from glass container.:
- Filtered through a 0.2 micron filter.:
Specification Test Results
About this item
Synonyms: Acetic acid ethyl ester
Boiling Pt:77.1 °C (1013 hPa)
Melting Pt:–83 °C
Density:0.902 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
Flash Pt:–4 °C
:Boiling Pt:
:Boiling Pt: 77.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –83 °C
:Flash Pt:
:Flash Pt: –4 °C
:M.W. 88.11 g/mol
Merck Index:13,03792
:Ethyl acetate
Formula:CH₃COOC₂H₅
MW:88.11 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00009171
CAS Number:141-78-6
EINECS:205-500-4
UN:1173
REACH:01-2119475103-46
:3
:II
:–4 °C
:GHS02
:GHS07
:M.O.C. Category N
Customers Who Bought This Also Bought