Water, B&J Brand™ for HPLC, for spectrophotometry, Burdick & Jackson™
Supplier: Burdick & Jackson
Certificates
About this item
Synonyms: Hydrogen oxide
Formula:H₂O
MW:18.02 g/mol
Boiling Pt:100 °C
Melting Pt:0 °C
Density:0.9971 g/cm³ (25 °C)
MDL Number:MFCD00011332
CAS Number:7732-18-5
Merck Index:12,10175
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 100 °C
:Melting Pt:
:Melting Pt: 0 °C
:Flash Pt:
:Flash Pt:
:M.W. 18.02 g/mol
:Water
EINECS:231-791-2
:100 °C
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Fluorescence (as quinine base):0.30 ppb max.
- UV Absorbance @ 400 nm:0.005 AU max.
- HPLC Gradient @ 200 nm:0.005 AU max.
- Microbial Contamination:Not detected
- Refractive Index (20ºC):1.3320 - 1.3440
- Residue:1 mg/L max.
- UV Absorbance @ 190 nm:0.01 AU max.
- UV Absorbance @ 200 nm:0.01 AU max.
- UV Absorbance @ 250 nm:0.005 AU max.
- UV Absorbance @ 300 nm:0.005 AU max.
- Burdick & Jackson™, Water, B&J Brand™ for HPLC, for spectrophotometry:
Specification Test Results
Customers Who Bought This Also Bought