Specifications
- Color:White
- Description:PTS Connect® CardioChek printer labels
- Cat. no.:77445-750
- Packaging:160 labels/roll
Specifications
About this item
For best results, the use of PTS Connect printer labels is recommended.
- Labels are sensitive to heat, moisture, and light
- Store labels at room temperature, in a cool, dry location
- Do not expose labels to high heat or direct light for an extended period of time
- Not for use with organic solvents
- One roll of 160 labels
The PTS Connect printer label roll is designed to be used with the PTS Connect printer. The printer labels have a sensor mark on the backing of each label for top-of-label alignment. Check to be sure the labels have this mark before loading them into the printer.