Dextrose, anhydrous 97.5-102.0% (by anhydrous basis), granular, GenAR® USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Corn sugar,Glucose,D-(+)-Dextrose,(2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal,Glucolin,Dextrose
CAS Number:50-99-7
:492-62-6
Formula:C₆H₁₂O₆
MW:180.16 g/mol
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
MDL Number:MFCD00148912
Merck Index:14,04459
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97,5-102,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 146 °C
:Flash Pt:
:Flash Pt:
:M.W. 180.16 g/mol
:Dextrose
Storage Temperature:Ambient
EINECS:200-075-1
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Identification C: Water (H₂O):≤1.0 %
- USP - Assay (Anhydrous basis):97.5 - 102.0 %
- USP - Related substances: Maltose and isomaltose:≤0.4 %
- USP - Dextrin:Passes Test
- USP - Related substances: Maltotriose:≤0.2 %
- USP - Related substances: Fructose:≤0.15 %
- USP - Related substances: Unspecified impurities, each:≤0.10 %
- USP - Related substances: Total Impurities:≤0.5 %
- USP - Color and Clarity of Solution:Passes Test
- USP - Soluble Starch, Sulfites:≤15 ppm
- USP - Conductivity (uS cm-1):≤20 µS/cm
- EP/BP - Identification A (Specific Rotation (+)):52.5 - 53.3 Degree
- EP/BP - Identification B:Passes Test
- EP/BP - Identification E:Passes Test
- EP/BP - Assay (C₆H₁₂O₆) (anhydrous substance):97.5 - 102.0 %
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Conductivity, uS cm-1:≤20
- EP/BP - Related Substances: sum of impurities A and B:≤0.4 %
- EP/BP - Related Substances: impurity C:≤0.2 %
- EP/BP - Related Substances: impurity D:≤0.15 %
- EP/BP - Related Substances: unspecified impurities, each:≤0.10 %
- EP/BP - Related Substances: total impurities:≤0.5 %
- EP/BP - Dextrins:Passes Test
- EP/BP - Soluble starch, sulfite:≤15 ppm
- EP/BP - Water (H₂O):≤1.0 %
- Endotoxin Concentration (EU/g):≤5.0
- JP - Identification 1:Passes Test
- JP - Identification 2:Passes Test
- JP - Water (H₂O):≤1.0 %
- JP - Assay (Anhydrous basis):97.5 - 102.0 %
- JP - Maltose and isomaltose:≤0.4 %
- JP - Maltotriose:≤0.2 %
- JP - Fructose:≤0.15 %
- JP - Unspecified Impurities, each:≤0.10 %
- JP - Total Impurities:≤0.5 %
- JP - Color and Clarity of Solution:Passes Test
- JP - Soluble Starch, Sulfites:≤15 ppm
- JP - Conductivity (uS cm-1):≤20 µS/cm
- JP - Dextrin:Passes Test
- JP - Heavy Metals (as Pb):≤4 ppm
- Appearance (white, granular powder):
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental impurities for this product is available on the associated Product Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number7730 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
Customers Who Bought This Also Bought