N,N-Dimethylacetamide ≥99.0% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog #: CAMK540708
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 127-19-5
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. no.:CAMK540708
- Supplier No.:5407-08
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CH₃CON(CH₃)₂) (by GC):≥99.0 %
- ACS - Residue after Evaporation:≤5 ppm
- ACS - Water (H₂O)(by Karl Fischer titrn):≤0.03 %
- Optical Absorbance (1-cm path vs water) - 270 nm:≤1.00
- Optical Absorbance (1-cm path vs water) - 280 nm:≤0.30
- Optical Absorbance (1-cm path vs water) - 290 nm:≤0.15
- Optical Absorbance (1-cm path vs water) - 310 nm:≤0.05
- Optical Absorbance (1-cm path vs water) - 320 nm:≤0.03
- Optical Absorbance (1-cm path vs water) - 360 - 400 nm:≤0.01
- Filtered through a 0.2 micron filter.:
- Appearance (clear, colorless liquid):
Specification Test Results
About this item
Synonyms: Acetic acid dimethylamide
Boiling Pt:166.1 °C (1013 hPa)
Melting Pt:–20 °C
Density:0.937 g/cm³ (20 °C)
ADR:6.1,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
Flash Pt:70 °C
:Boiling Pt:
:Boiling Pt: 166.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –20 °C
:Flash Pt:
:Flash Pt: 70 °C
:M.W. 87.12 g/mol
Merck Index:13,03254
:N,N-Dimethylacetamide
Formula:CH₃CON(CH₃)₂
MW:87.12 g/mol
MDL Number:MFCD00008686
CAS Number:127-19-5
EINECS:204-826-4
UN:2810
:6.1
:III
:GHS08
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
Customers Who Bought This Also Bought