Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, Biotech Reagent, J.T.Baker®
Catalog #: CAJTF781-5
Supplier: VWR International
CAS Number: 3483-12-3
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:1 kg
- Supplier No.:F781-05
Specifications
Specification Test Results
- GMP Manufactured Product:
- Keep refrigerated between 2-8°C (36-46°F) and store under nitrogen.:
- CAUTION: For Manufacturing, processing or repackaging:
- Assay (HSCH₂(CHOH)₂CH₂SH):≥99.0 %
- Appearance:Passes Test
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 280 nm:≤0.060
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:≤0.400
- Infrared Spectrum:Passes Test
- J.T.Baker®, Dithiothreitol (DTT, Cleland's reagent) ≥99.0%, Biotech Reagent:
Specification Test Results
About this item
Synonyms: cis-2,3-Dihydroxy-1,4-butanedithiol,(R*,R*)-1,4-Dimercapto-2,3-butandiol,DTT,DL-1
Boiling Pt:∼125 °C (1013 hPa)
Melting Pt:42…44 °C
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = R
Flash Pt:>110 °C
:Boiling Pt:
:Boiling Pt: ∼125 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 42…44 °C
:Flash Pt:
:Flash Pt: >110 °C
:M.W. 154.25 g/mol
:Dithiothreitol (DTT, Cleland's reagent)
CAS Number:3483-12-3
EINECS:222-468-7
:∼125 °C (1013 hPa)
:>110 °C
:GHS07
Formula:C₄H₁₀O₂S₂
Storage Temperature:Refrigerator
:M.O.C. Category R
:27565-41-9
MW:154.25 g/mol
MDL Number:MFCD00004877
Customers Who Bought This Also Bought