N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BAKER BIO-ANALYZED® for biotechnology, J.T.Baker®
Catalog #: CAJT9344-13
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 68-12-2
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. no.:CAJT9344-13
- Supplier No.:9344-13
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (HCON(CH₃)₂) (by GC, corrected for water):≥99.8 %
- Appearance:Passes Test
- Color (APHA):≤15
- Residue after Evaporation:≤5 ppm
- Titrable Acid (meq/g):≤0.0005
- Titrable Base (meq/g):≤0.003
- Water (H₂O)(by Karl Fischer titrn):≤400 ppm
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:≤0.010
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 340 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 310 nm:≤0.05
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 295 nm:≤0.10
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:≤0.30
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 270 nm:≤1.00
- Ultraviolet Absorbance (1.00-cm cell vs. water) - Amines (as dimethylamine):≤5 ppm
Specification Test Results
About this item
Synonyms: Formic acid dimethylamide,DMFA,Formyldimethylamine,N-Formyldimethylamine,Dimethylformamide,NSC 5356
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Management of Change (MOC) category = N
Flash Pt:57.5 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 153 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –61 °C
:Flash Pt:
:Flash Pt: 57.5 °C (closed cup)
:M.W. 73.09 g/mol
Merck Index:13,03269
Formula:HCON(CH₃)₂
MW:73.09 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003284
CAS Number:68-12-2
EINECS:200-679-5
UN:2265
REACH:01-2119475605-32
:3
:III
:57.5 °C
:GHS02
:GHS08
:GHS07
:M.O.C. Category N
Customers Who Bought This Also Bought