Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: VWR International
MOC: Non-Regulated
About this item
Synonyms: Methylene chloride
Formula:CH₂Cl₂
MW:84.93 g/mol
Boiling Pt:39.8 °C (1013 hPa)
Melting Pt:–95 °C
Density:1.322 g/cm³ (20 °C)
MDL Number:MFCD00000881
CAS Number:75-09-2
ADR:6.1,III
Merck Index:12,06140
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt: 39.8 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -95 °C|Melting Pt: –95 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.93 g/mol
:Dichloromethane
EINECS:200-838-9
UN:1593
:6.1
:III
:GHS08
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (CH₂Cl₂) (by GC, corrected for water):≥99.5 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.0020 %
- Titrable Acid (µeq/g):≤0.3
- Free Halogens:Passes Test
- Water (by KF, coulometric):≤0.02 %
- Product Information (not specifications)::
- Appearance (Clear, colorless liquid):
- Boiling Point (typical)................. 39.5 °C.:
- Density (g/mL) at 25 °C (typical)........ 1.318:
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Dichloromethane ≥99.5% (by GC, corrected for water content), BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number9324 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
JTBaker_9324_Datasheet.pdf
Datasheet
Customers Who Bought This Also Bought