Dichloromethane, low water content ≥99.5% stabilized, BakerDRY™, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Methylene chloride
Formula:CH₂Cl₂
MW:84.93 g/mol
Boiling Pt:39.5 °C
Melting Pt:–95 °C
Density:1.318 g/cm³
MDL Number:MFCD00000881
CAS Number:75-09-2
ADR:6.1,III
Merck Index:12,06140
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = N
:Stabilised with cyclohexene
:Boiling Pt:
:Boiling Pt: 39.5 °C
:Melting Pt:
:Melting Pt: –95 °C
:Flash Pt:
:Flash Pt:
:M.W. 84.93 g/mol
:Dichloromethane
EINECS:200-838-9
:39.5 °C
UN:1593
:6.1
:III
:GHS08
:GHS07
Storage Temperature:Ambient
:Cyclohexen
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (by GC, corrected for water, exclusive of preservative):≥99.5 %
- Appearance:Passes Test
- ACS - Color (APHA):≤10
- ACS - Free Halogens:Passes Test
- ACS - Preservative (cyclohexene), For Information Only:
- ACS - Residue after Evaporation:≤2 ppm
- ACS - Titrable Acid (µeq/g):≤0.2
- ACS - Water (by KF, coulometric):≤60 ppm
- Product Information (not specifications)::
- Boiling Point (typical)................. 39.5 °C.:
- Density (g/mL) at 25 °C (typical)........1.318:
Specification Test Results
Related Information
Certificates
Search by Lot Number9295 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
BakerDry Grade Definition.png
Datasheet
Customers Who Bought This Also Bought