Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial, J.T.Baker®
Supplier: VWR International
Certificates
About this item
Synonyms: Methyl sulfoxide,Methyl sulphoxide
Formula:(CH₃)₂SO
MW:78.14 g/mol
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.45 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:89 °C (closed cup)
MDL Number:MFCD00002089
CAS Number:67-68-5
Merck Index:13,03285
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 189 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.5 °C|Melting Pt: 18.45 °C
:Flash Pt:
:Flash Pt: 89 °C (closed cup)
:M.W. 78.14 g/mol
EINECS:200-664-3
:89 °C
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets U.S.P Requirements:
- Meets E.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Specific Gravity at 25°/25°C:1.095 - 1.101
- USP - Refractive Index, n²⁵^D:1.4755 - 1.4775
- USP - Acidity:Passes Test
- USP - Water (H₂O):≤ 0.1 %
- USP - Absorbance - 275 nm:≤ 0.20
- USP - Absorbance - 270-350nm (smooth with no maxima):Passes Test
- USP - Absorbance Ratio - 285 nm/275 nm:≤ 0.65
- USP - Absorbance Ratio - 295 nm/275 nm:≤ 0.45
- USP - Nonvolatile Residue:≤ 0.01 %
- USP - Related Compounds:≤ 0.1 %
- USP - Assay ((CH₃)₂SO):≥ 99.9 %
- EP - Identification:Passes Test
- EP - Acidity:Passes Test
- EP - Relative Density:1.100 - 1.104
- EP - Refractive Index:1.478 - 1.480
- EP - Freezing Point:≥ 18.3 °C
- EP - Absorbance - 275 nm:≤ 0.30
- EP - Absorbance - 285 nm:≤ 0.20
- EP - Absorbance - 295 nm:≤ 0.20
- EP - Absorbance - 270-350nm (smooth with no maxima):Passes Test
- EP - Related Substances: Unspecified impurities, Each:≤ 0.10 %
- EP - Related Substances-Total Impurities:≤ 0.15 %
- EP - Water (H₂O):≤ 0.2 %
- Water (H₂O), For Information Only:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
- J.T.Baker®, Dimethyl sulfoxide ≥99.9% USP, Multi-Compendial:
Specification Test Results
Related Information
Certificates
Search by Lot Number9033 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
sds_BAKR9033
SDS
spec_JT9033-07 spec_JT9033-33
Product specification
Customers Who Bought This Also Bought