1,4-Dioxane ≥99.0%, GR ACS, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Glycolethylether,1,4-Dioxacyclohexane,Dioxane,p-Dioxane,1
Formula:C₄H₈O₂
MW:88.11 g/mol
Boiling Pt:101.1 °C (1013 hPa)
Melting Pt:11.8 °C
Density:1.03 g/cm³ (20 °C)
Flash Pt:12 °C
MDL Number:MFCD00006571
CAS Number:123-91-1
ADR:3,II
Merck Index:13,03330
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Boiling Pt:
:Boiling Pt: 101.1 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 11.8 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 88.11 g/mol
:1,4-Dioxane
EINECS:204-661-8
UN:1165
:3
:II
:GHS02
:GHS08
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Expiration date:Exp 60 months from mfg date
- Assay (GC):99.0% min
- Carbonyl (as HCHO):0.01% max
- Color (APHA):20 max
- Freezing point:11.0°C min
- Identity (IR-spectrum):Conforms
- Peroxide (as H2O2):0.005% max
- Residue after evaporation:0.005% max
- Titrable acid:0.0016meq/g max
- Water (H2O):0.05% max
- Supelco®, 1,4-Dioxane ≥99.0%, GR ACS:
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Start and End with Quality Product Portfolio Brochure
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought