N,N-Dimethylformamide, low water content ≥99.5% ACS, VWR Chemicals BDH®
Catalog #: BDH1119-19L
Supplier: VWR International
CAS Number: 68-12-2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Steel can
- Size:19 L
- Cat. no.:BDH1119-19L
- Supplier No.:PL076AA-19
Specifications
Specification Test Results
- Appearance:Clear, colorless liquid (can be slightly yellow)
- Water (K.F.):Less than 300 ppm
- GC Purity:Greater than 99.5%
- Color (APHA):Less than 15
- Amines [as dimethylamine (DMA)]:Less than 10 ppm
- VWR Chemicals BDH®, N,N-Dimethylformamide, low water content ≥99.5% ACS:
Specification Test Results
About this item
Synonyms: Formic acid dimethylamide,DMFA,Formyldimethylamine,N-Formyldimethylamine,Dimethylformamide,NSC 5356
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
Flash Pt:57.5 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 153 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -61 °C|Melting Pt: –61 °C
:Flash Pt:
:Flash Pt: 57.5 °C (closed cup)
:M.W. 73.09 g/mol
Merck Index:13,03269
Formula:HCON(CH₃)₂
MW:73.09 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00003284
CAS Number:68-12-2
EINECS:200-679-5
UN:2265
REACH:01-2119475605-32
:3
:III
:57.5 °C
:GHS02
:GHS08
:GHS07
Customers Who Bought This Also Bought