Dimethyl sulfoxide ≥99.9% ACS, VWR Chemicals BDH®
Supplier: VWR International
MOC: Non-Regulated
About this item
Synonyms: Methyl sulfoxide,Methyl sulphoxide
Formula:(CH₃)₂SO
MW:78.14 g/mol
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C
MDL Number:MFCD00002089
CAS Number:67-68-5
Merck Index:13,03285
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Boiling Pt:
:Boiling Pt: 189 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 18.5 °C
:Flash Pt:
:Flash Pt: 87 °C
:M.W. 78.14 g/mol
Storage Temperature:Ambient
EINECS:200-664-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:Clear, colorless liquid
- Water (K.F.):Less than 0.10%
- Residue:Less than 0.01% (0.0010g/91mL)
- GC Purity:Greater than 99.9%
- Color (APHA):Less than 10
- Titrable acid:Less than 1µeq/g (Not more than 5.0mL of of 0.01N sodium hydroxide)
- VWR Chemicals BDH®, Dimethyl sulfoxide ≥99.9% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
BDH1115-1LP_BSETSE BDH1115-1LP_P BDH1115-4LP_BSETSE BDH1115-4LP_P
BSE/TSE Certificates
spec_BDH1115
Product specification
Customers Who Bought This Also Bought