Chloroform ≥99.8% (by GC, corrected for water content) stabilized, ChromAR® ACS for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Methane, trichloro-,Trichloroform
Formula:CHCl₃
MW:119.38 g/mol
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
MDL Number:MFCD00000826
CAS Number:67-66-3
ADR:6.1,III
Merck Index:13,02160
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Management of Change (MOC) category = N
:Stabilised with amylene
:Boiling Pt:
:Boiling Pt: 61.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –63 °C
:Flash Pt:
:Flash Pt:
:M.W. 119.38 g/mol
EINECS:200-663-8
UN:1888
REACH:05-2114614906-45
:6.1
:III
:GHS06
:GHS08
Storage Temperature:Ambient
:Amylen
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CHCl₃) (by GC, corrected for water):≥99.8 %
- ACS - Acetone, Aldehydes (as (CH₃)₂CO):Passes Test
- ACS - Acid and Chloride:Passes Test
- ACS - Color (APHA):≤10
- ACS - Free Chlorine:Passes Test
- ACS - Lead (Pb):≤0.050 ppm
- ACS - Residue after Evaporation:≤0.0003 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Suitability for Use in Dithizone Tests:Passes Test
- ACS - Water (H₂O):≤0.02 %
- ACS - Ultraviolet Absorbance (1.00-cm cell vs. water)::Passes Test
- ACS UV Absorbance - 245 nm:≤1.00
- ACS UV Absorbance - 254 nm:≤0.15
- ACS UV Absorbance - 255 nm:≤0.25
- ACS UV Absorbance - 260 nm:≤0.15
- ACS UV Absorbance - 265 nm:≤0.05
- ACS UV Absorbance - 270 nm:≤0.05
- ACS Absorbance 280 nm:≤0.01
- ACS Absorbance 300 nm:≤0.01
- ACS Absorbance - 350 nm:≤0.005
- ACS Absorbance - 350 - 400 nm:≤0.01
- ACS - Absorbance:Passes Test
- Appearance (clear, colorless liquid):
- Macron Fine Chemicals™, Chloroform ≥99.8% (by GC, corrected for water content) stabilized, ChromAR® ACS for liquid chromatography, for UV spectrophotometry:
Specification Test Results
Related Information
Certificates
Search by Lot Number4443 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_MK444306
Product specification
Customers Who Bought This Also Bought