Calcium phosphate tribasic, powder NF, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Hydroxylapatite,tri-Calcium phosphate,HAp,Calcium Hydroxide Phosphate,Calcium hydroxyphosphate
Formula:Ca₅HO₁₃P₃
MW:502.306 g/mol
Density:3.14 g/cm³
MDL Number:MFCD00010904
CAS Number:12167-74-7
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 502.306 g/mol
:Calcium phosphate tribasic
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets F.C.C. Requirements:
- GMP Manufactured Product:
- Meets N.F. Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- NF - Assay (Ca):34.0 - 40.0 %
- NF - Acid-Insoluble Substances:≤ 0.2 %
- NF - Arsenic (As):≤ 3 mg/kg
- NF - Barium (Ba):Passes Test
- NF - Carbonate (CO₃):Passes Test
- NF - Chloride (Cl):≤ 0.14 %
- NF - Dibasic Salt and Calcium Oxide (ml):13.0 - 14.3
- NF - Fluoride (F):≤ 0.0075 %
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Loss on Ignition:≤ 8.0 %
- NF - Nitrate (NO₃):Passes Test
- NF - Sulfate (SO₄):≤ 0.8 %
- NF - Water-Soluble Substances:≤ 0.5 %
- FCC - Assay (Ca):34.0 - 40.0 %
- FCC - Arsenic (As):≤ 3 mg/kg
- FCC - Fluoride (F):≤ 0.0075 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Lead (Pb):≤ 2 mg/kg
- FCC - Loss on Ignition:≤ 8.0 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Macron Fine Chemicals™, Calcium phosphate tribasic, powder NF, FCC:
Specification Test Results
Related Information
Certificates
Search by Lot Number4280 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR4280
SDS
Customers Who Bought This Also Bought