Chloroform ≥99.8% (by GC, corrected for water content) stabilized, BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Methane, trichloro-,Trichloroform
Formula:CHCl₃
MW:119.38 g/mol
Boiling Pt:61.7 °C (1013 hPa)
Melting Pt:–63 °C
Density:1.4832 g/cm³ (20 °C)
MDL Number:MFCD00000826
CAS Number:67-66-3
ADR:6.1,III
Merck Index:13,02160
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Management of Change (MOC) category = N
:Stabilised with ethanol ∼0.75%
:Boiling Pt:
:Boiling Pt: 61.7 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -63 °C|Melting Pt: –63 °C
:Flash Pt:
:Flash Pt:
:M.W. 119.38 g/mol
EINECS:200-663-8
UN:1888
REACH:05-2114614906-45
:6.1
:III
:GHS07
:GHS08
Storage Temperature:Ambient
:EtOH
:∼0.75%
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-280 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.15
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤245
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.2 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (CHCl₃) (by GC, exclusive of preservative, corrected for water):≥99.8 %
- Chloride (Cl):≤10 ppm
- Residue after Evaporation:≤2.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Titrable Acid (µeq/g):≤0.5
- Water (by KF, coulometric):≤100 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number9175 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
JTBaker_9175_Datasheet.pdf
Datasheet
sds_BAKR9175
SDS
Customers Who Bought This Also Bought