Calcium chloride dihydrate 99.0-107.0%, CalPlus™ USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Calcium Chloride Dihydrate
Formula:CaCl₂·2H₂O
MW:147.01 g/mol
Boiling Pt:1600 °C (1013 hPa)
Melting Pt:176 °C
Density:1.85 g/cm³ (20 °C)
MDL Number:MFCD00149613
CAS Number:10035-04-8
Merck Index:13,01660
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-107,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 1600 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 176 °C
:Flash Pt:
:Flash Pt:
:M.W. 147.01 g/mol
:Calcium chloride dihydrate
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets F.C.C. Requirements:
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets J.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Food GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- USP - Assay (CaCl₂·2H₂O):99.0 - 107.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - pH of 5% Solution at 25°C:4.5 - 9.2
- USP - Iron, Aluminum, and Phosphate:Passes Test
- USP - Magnesium and Alkali Salts:≤ 1.0 %
- USP - Aluminum (Al)(µg/g):≤ 1
- Endotoxin Concentration (EU/g):≤ 10
- FCC - Assay (CaCl₂·2H₂O):99.0 - 107.0 %
- FCC - Identification A:Passes Test
- FCC - Identification B:Passes Test
- FCC - Arsenic (As):≤ 3 mg/kg
- FCC - Magnesium and Alkali Salts:≤ 1.0 %
- FCC - Lead (Pb):≤ 5 mg/kg
- FCC - Fluoride (F):≤ 0.004 %
- Appearance (White crystals or granules):Passes Test
- EP/BP - Assay (CaCl₂·2H₂O):97.0 - 103.0 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Identification C:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Sulfate (SO₄):≤ 300 ppm
- EP - Aluminum (Al):≤ 1 ppm
- EP/BP - Iron (Fe):≤ 10 ppm
- EP/BP - Magnesium and Alkali Metals:≤ 0.5 %
- JP - Assay (CaCl₂·2H₂O):96.7 - 103.3 %
- JP - Identification:Passes Test
- JP - pH (1 in 20):4.5 - 9.2
- JP - Heavy Metals (as Pb):≤ 10 ppm
- JP - Clarity and Color of Solution:Passes Test
- JP - Sulfate (SO₄):≤ 0.024 %
- JP - Hypochlorite:Passes Test
- JP - Iron, Aluminum or Phosphate:Passes Test
- JP - Barium (Ba):Passes Test
- JP - Arsenic (As):≤ 2 ppm
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Suitable for use in hemodialysis.:
Specification Test Results
Related Information
Certificates
Search by Lot Number1336 Product Specifications
Additional Product Details
sds_BAKR1336
SDS
spec_JT1336-1 spec_JT1336-5 spec_JT1336-7 spec_JT1336-8 spec_JT1336-9
Product specification
More
Customers Who Bought This Also Bought