Citric acid monohydrate 99.5-100.5% (by anhydrous basis) USP, FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Hydroxytricarballylic acid,2-Hydroxy-1,2
Formula:C₆H₈O₇.H₂O
MW:210.14 g/mol
Boiling Pt:280…285 °C (1013 hPa)
Melting Pt:135…152 °C
Density:1.54 g/cm³
Flash Pt:173.9 °C (closed cup)
MDL Number:MFCD00149972
CAS Number:5949-29-1
Merck Index:13,02350
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 280…285 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 135 to 152 °C|Melting Pt: 135…152 °C
:Flash Pt:
:Flash Pt: 173.9 °C (closed cup)
:M.W. 210.14 g/mol
:Citric acid monohydrate
EINECS:201-069-1
:280…285 °C (1013 hPa)
:173.9 °C
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Food GMP Manufactured Product:
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets F.C.C. Requirements:
- Preserve in Tight Containers:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- USP - Assay (as C₆H₈O₇) (calculated on anhydrous basis):99.5 - 100.5 %
- USP - Clarity of Solution:Passes Test
- USP - Color of Solution:Passes Test
- USP - Identification (USP):Passes Test
- USP - Water (H₂O) (U.S.P.):7.5 - 9.0 %
- USP - Residue on Ignition:≤0.05 %
- USP - Oxalic Acid (USP):≤0.036 %
- USP - Sulfate (SO₄):≤0.015 %
- USP - Readily Carbonizable Substances:Passes Test
- FCC - Assay (as C₆H₈O₇) (calculated on anhydrous basis):99.5 - 100.5 %
- FCC - Identification (FCC):Passes Test
- FCC - Water (H₂O) (F.C.C.):≤8.8 %
- FCC - Residue on Ignition:≤0.05 %
- FCC - Oxalate (C₂O₄) (FCC):≤100 mg/kg
- FCC - Readily Carbonizable Substances:Passes Test
- FCC - Lead (Pb):≤0.5 mg/kg
- Appearance (white, crystalline powder):Passes Test
- Must be subjected to further processing during the preparation of:
- injectable dosage forms.:
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232) - Information on elemental impurities for:
- this product is available on the associated Product Regulatory Data:
- Sheet and elemental impurity profile report.:
- J.T.Baker®, Citric acid monohydrate 99.5-100.5% (by anhydrous basis) USP, FCC:
Specification Test Results
Related Information
Certificates
Search by Lot Number0119 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT0119-7
Product specification
Customers Who Bought This Also Bought