Evolution One™ Latex Gloves, Microflex®, Ansell
Supplier: Avantor
CE Compliant
Certificates
About this item
These ambidextrous latex gloves have added coating that provides super antisticking properties.
- Ambidextrous
- Interior polymer coating for superior antisticking properties in all conditions
- Fully textured, beaded cuff, powder-free
- Thickness: 8.7 mil (finger)
These gloves have an interior polymer coating that provides superior antisticking properties, resulting in highly effective donning in wet, damp, or dry conditions.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Specifications
- Catalog No:
- 32916-536
- 32916-534
- 32916-530
- CA32916-538
- 32916-532
- CA32916-536
- CA32916-534
- CA32916-532
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- Finger thickness:
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- 8.7 mil
- Palm thickness:
- 6.7 mil
- 6.7 mil
- 6.7 mil
- 6.7 mil
- 6.7 mil
- 6.7 mil
- 6.7 mil
- 6.7 mil