Diamond Grip Plus™ Latex Gloves, Microflex®, Ansell
Supplier: Microflex
CE Compliant
Certificates
About this item
Thick latex gloves are textured for superior grip and provide excellent protection from rips and tears.
- Ambidextrous
- Beaded cuff
Gloves are manufactured with integrated polymer technologies for powder-free donning and product consistency. Fully textured for a secure grip in wet or dry conditions.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Specifications
- Catalog No:
- CA32916-636
- CA32916-634
- CA32916-632
- CA32916-640
- CA32916-638
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Finger thickness:
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- Palm thickness:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil