Diamond Grip™ Latex Gloves, Microflex®, Ansell
Supplier: Microflex
CE Compliant
Certificates
About this item
Thick latex gloves are textured for superior grip and provide excellent protection from rips and tears.
- Thick textured fingers for gripping in wet and dry conditions
- Powder-free
- Fully chlorinated
- Beaded cuff
- Thickness: 9.1 mil (finger)
These gloves are thicker than typical latex gloves for enhanced protection. They are thickest at the fingertips, providing superior protection from rips and tears.
Caution: This product contains natural rubber latex which may cause allergic reactions. Safe use of this glove by or on latex-sensitized individuals has not been established.
Specifications
- Catalog No:
- CA32916-502
- CA32916-500
- CA32916-506
- CA32916-503
- CA32916-498
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- Finger thickness:
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- 9.1 mil
- Palm thickness:
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil
- 6.3 mil