Chloroform ≥99.8% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Supplier: Burdick & Jackson
Certificates
About this item
Synonyms: Methane, trichloro-,Trichloroform
Formula:CHCl₃
MW:119.38 g/mol
Boiling Pt:62 °C
Melting Pt:-64 °C
Density:1.49 g/cm³ (20 °C)
Flash Pt:Does not flash
MDL Number:MFCD00000826
CAS Number:67-66-3
ADR:6.1,III
Merck Index:13,02160
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Stabilised with amylene
:Boiling Pt:
:Boiling Pt: 62 °C
:Melting Pt:
:Melting Pt: -63 °C|Melting Pt: -64 °C
:Flash Pt:
:Flash Pt: Does not flash
:M.W. 119.38 g/mol
EINECS:200-663-8
:62 °C
UN:1888
REACH:05-2114614906-45
:6.1
:III
:Does not flash
:GHS08
:GHS07
Storage Temperature:Ambient
:Amylen
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:Passes test
- Assay by GC Analysis:99.8 % min.
- Color (APHA):10 max.
- Free Chlorine:Passes test
- Lead:0.05 ppm max.
- Residue after Evaporation:0.001 % max.
- Water by Karl Fischer Titration:0.02 % max.
- Chloroform ≥99.8% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use:
Specification Test Results
Related Information
Certificates
Search by Lot NumberHB Honeywell B
Datasheet
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought