Chlorobenzene ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry, Burdick & Jackson™
Catalog #: CABJ043-4
Supplier: Burdick & Jackson
CAS Number: 108-90-7
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:4 L
- Cat. no.:CABJ043-4
- Supplier No.:043-4
Specifications
Specification Test Results
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.5242 - 1.5256
- UV Absorbance @ 300 nm:0.05 AU max.
- UV Absorbance @ 325 nm:0.04 AU max.
- UV Absorbance @ 350 nm:0.02 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:287 nm max.
- Water by Karl Fischer Titration:0.03 % max.
- Burdick & Jackson™, Chlorobenzene ≥99.9%, B&J Brand™ for HPLC, for gas chromatography, for spectrophotometry:
Specification Test Results
About this item
Synonyms: Phenylchloride
Boiling Pt:132 °C
Melting Pt:-46 °C
Density:1.1058 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
Flash Pt:28 °C
:Boiling Pt:
:Boiling Pt: 132 °C
:Melting Pt:
:Melting Pt: -45 °C|Melting Pt: -46 °C
:Flash Pt:
:Flash Pt: 28 °C
:M.W. N/A N/A
Merck Index:13,02139
Formula:C₆H₅Cl
MW:112.56 g/mol
MDL Number:MFCD00000530
CAS Number:108-90-7
EINECS:203-628-5
:132 °C
Storage Temperature:Ambient
UN:1134
REACH:01-2119432722-45
:3
:III
:28 °C
:GHS02
:GHS07
:GHS09
Customers Who Bought This Also Bought