Cyclohexane ≥99% ACS, VWR Chemicals BDH®
Supplier: VWR International
Certificates
About this item
Synonyms: Hexamethylene
Formula:C₆H₁₂
MW:84.16 g/mol
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
MDL Number:MFCD00003814
CAS Number:110-82-7
ADR:3,II
Merck Index:13,02752
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Boiling Pt:
:Boiling Pt: 81 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 7 °C
:Flash Pt:
:Flash Pt: –18 °C
:M.W. 84.16 g/mol
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
:3
:II
:–18 °C
:GHS02
:GHS08
:GHS07
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Appearance:Clear
- Water (K.F.):Less than 0.02%
- Residue:Less than 0.002% (0.0020 g/129 mL)
- GC Purity:Greater than 99.0%
- Color (APHA):Less than 10
- Substances darkened by sulfuric acid:Passes test (Neither the cyclohexane nor acid layer should be darkened.)
- VWR Chemicals BDH®, Cyclohexane ≥99% ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number12777743.pdf 12777745.pdf BDH1111-4LG_BSETSE
BSE/TSE Certificates
19-20 l Close Top Steel Can with Handle
Packaging Information
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
ghs_BDH1111
SDS
spec_BDH1111
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought