Xylene (mixture of isomers) ≥98.5% (by GC) contains ethylbenzene, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Dimethylbenzene
Formula:C₆H₄(CH₃)₂
MW:106.17 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
MDL Number:MFCD00077264
CAS Number:1330-20-7
ADR:3,III
Merck Index:12,10214
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5
:Boiling Pt:
:Boiling Pt: 140 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –34 °C
:Flash Pt:
:Flash Pt: 25 °C
:M.W. 106.17 g/mol
EINECS:215-535-7
:140
UN:1307
:3
:III
:25
:GHS02
:GHS07
:GHS08
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (m,p,o-xylenes and ethylbenzene) (by GC):≥ 98.5 %
- Ethylbenzene (C₆H₅C₂H₅):≤ 25 %
- Color (APHA):≤ 10
- Residue after Evaporation:≤ 0.0020 %
- Substances Darkened by H₂SO₄:Passes Test
- Sulfur Compounds (as S):≤ 0.0005 %
- Water (H₂O):≤ 0.02 %
- Particulate Contamination:Passes Test
- Date Received: __ __ /__ __ /__ __:
- Date Opened: __ __ /__ __ /__ __:
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Xylene (mixture of isomers) ≥98.5% (by GC) contains ethylbenzene, BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number9490 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR9490
SDS
spec_JT9490-1
Product specification
Customers Who Bought This Also Bought