Xylene (mixture of isomers) ACS for production, VWR Chemicals BDH®
Supplier: VWR International
MOC: Non-Regulated
About this item
Synonyms: Dimethylbenzene
Formula:C₆H₄(CH₃)₂
MW:106.17 g/mol
Boiling Pt:140 °C (1013 hPa)
Melting Pt:–34 °C
Density:0.86…0.88 g/cm³ (20 °C)
Flash Pt:25 °C
MDL Number:MFCD00077264
CAS Number:1330-20-7
ADR:3,III
Merck Index:12,10214
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt: 140 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -34 to 34 °C|Melting Pt: –34 °C
:Flash Pt:
:Flash Pt: 25 °C
:M.W. 106.17 g/mol
EINECS:215-535-7
UN:1307
:3
:III
:GHS02
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay by GC (Ethylbenzene):25.0% Max.
- GC Purity (Xylenes + Ethylbenzene):98.5% Min.
- Color (APHA):10 APHA Max.
- Residue upon Evaporation:0.002% Max.
- Substances Darkened by Sulfuric Acid:Passes Test
- Sulfur Compounds (as S):0.003% Max.
- Water:0.05% Max.
- VWR Chemicals BDH®, Xylene (mixture of isomers) ACS for production:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
spec_BDH2000-55GLM spec_BDH2000-5GLMS
Product specification
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought