KONTES® Reaction Beaker, Glass Jacketed, Kimble Chase
Catalog #:
Supplier: DWK Life Sciences (KIMBLE)
Supplier #:
About this item
Useful in biological and chemical reactions including titrations, enzyme digestion and synthetic organic reactions.
- Jacket has #2 inlet/outlet serrated hose connections
- Conical beaker bottom accepts egg-shaped PTFE magnetic stir bar for optimum agitation without spin-out
- Open top provides good access to concentrated residual samples
Borosilicate glass construction designed for temperature controlled reactions.
Certifications: Design meets ASTM Specification E960, Type I requirements. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought