Bromothymol blue sodium salt in aqueous solution, J.T.Baker®
Catalog #: CAJTD472-7
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 34722-90-2
MOC: Non-RegulatedCertificates
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:500 ml
- Cat. no.:CAJTD472-7
- Supplier No.:D472-07
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Visual Transition Intervals - pH…7.6 (Blue):Passes Test
- Visual Transition Intervals - pH…6.7 (Green):Passes Test
- Visual Transition Intervals - pH…6.0 (Yellow):Passes Test
- Alkali Consumption:Passes Test
- in aqueous solution:
Specification Test Results
About this item
Synonyms: Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulphonate,Bromothymol blue water soluble,Sodium α-(3-bromo-5-isopropyl-4-oxo-2-methyl-2,5-cyclohexadienylidene)-2-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)toluenesulfonate,Sodium 2-bromo-4-(3-(3-bromo-4-hydroxy-5-isopropyl-2-methylphenyl)-1,1-dioxido-3H-benzo[c][1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: Min. 200 °C
:Flash Pt:
:Flash Pt:
:M.W. 646.37 g/mol
Formula:C₂₇H₂₇Br₂NaO₅S
MW:646.37 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00077263
CAS Number:34722-90-2
EINECS:252-169-7
:M.O.C. Category N
Customers Who Bought This Also Bought