To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:IGFBP4
- Clonality:Polyclonal
- Gene ID:3487
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Rat,Mouse
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Western blot:Yes
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:26
- Storage temperature:Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
- Concentration:1 µg/µl
- Purification:Purified by protein A
- Packaging:Vial
Specifications
About this item
Insulin like growth factor binding protein 4 (IGFBP4) is produced from a DNA sequence encoding the human IGFBP4 protein. Mature human IGFBP4 has a calculated molecular mass of approximately 26 kDa. Due to glycosylation, the recombinant protein migrates as a 32 kDa and 25 kDa protein under reducing and non reducing conditions, respectively. Human IGFBP4 has a potential N-linked glycosylation site and shares approximately 90% amino acid sequence identity with both mouse and rat IGFBP4. IGFBP4 is a member of the superfamily of insulin-like growth factor (IGF) binding proteins which include six high affinity IGF binding proteins (IGFBP) and at least four low affinity binding proteins referred to as IGFBP related proteins (IGFBPrP). IGFBP4 functions as an inhibitor of IGF action and its main function may be to protect cells from overstimulation by IGFs or to allow activation of alternate transmembrane signaling pathways that are inhibited by IGF exposure. IGFBP4 is expressed in multiple tissues including adrenal, testis, spleen, heart, lung, kidney, liver, stomach, hypothalamus, and brain cortex.
Type: Primary
Antigen: IGFBP4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Mouse, Rat