Benzyl alcohol 98.0-100.5% NF, Multi-Compendial low in peroxide, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Phenylmethanol
Formula:C₆H₅CH₂OH
MW:108.14 g/mol
Boiling Pt:205 °C (1013 hPa)
Melting Pt:–15.3 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:101 °C
MDL Number:MFCD00004599
CAS Number:100-51-6
Merck Index:13,01126
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 205 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –15.3 °C
:Flash Pt:
:Flash Pt: 101 °C
:M.W. 108.14 g/mol
EINECS:202-859-9
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets J.P. Chemical Specifications:
- Meets N.F. Requirements:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Assay:98.0 - 100.5 %
- NF - Clarity of Solution:Passes Test
- NF - Color of Solution:Passes Test
- NF - Identification:Passes Test
- NF - Peroxide Value:≤5
- NF - Related Substances - Benzaldehyde:≤0.05 %
- NF - Related Substances - Cyclohexylmethanol:≤0.10 %
- NF - Related Substances - Total Other Peaks <RT of Benzyl Alcohol:≤0.02 %
- NF - Related Substances - Total other peaks >RT of benzyl alcohol:≤0.2 %
- NF - Refractive Index, n²ºD:1.538 - 1.541
- NF - Acidity:Passes Test
- NF - Residue on Evaporation:≤0.05 %
- Solubility:Passes Test
- EP/BP - Assay:98.0 - 100.5 %
- EP/BP - Identification:Passes Test
- EP/BP - Acidity:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Refractive Index, n²ºD:1.538 - 1.541
- EP/BP - Peroxide Value:≤5
- EP/BP - Residue on Evaporation:≤0.05 %
- EP/BP - Related Substances - Benzaldehyde:≤0.05 %
- EP/BP - Related Substances - Cyclohexylmethanol:≤0.10 %
- EP/BP - Related Substances - Total Other Peaks <RT of Benzyl Alcohol:≤0.02 %
- EP/BP - Related Substances - Total other peaks >RT of benzyl alcohol:≤0.2 %
- JP - Assay:98.0 - 100.5 %
- JP - Identification:Passes Test
- JP - Refractive Index, n²ºD:1.538 - 1.541
- JP - Specific Gravity at 20°/20°C:1.043 - 1.049
- JP - Clarity and Color of Solution:Passes Test
- JP - Acidity:Passes Test
- JP - Related Substances - Benzaldehyde:≤0.05 %
- JP - Related Substances - Cyclohexylmethanol:≤0.10 %
- JP - Related Substances - Total Other Peaks <RT of Benzyl Alcohol:≤0.02 %
- JP - Related Substances - Total other peaks >RT of benzyl alcohol:≤0.2 %
- JP - Peroxide Value:≤5
- JP - Residue on Evaporation:≤0.05 %
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Suitable for use in the manufacture of parenteral dosage forms.:
- J.T.Baker®, Benzyl alcohol 98.0-100.5% NF, Multi-Compendial low in peroxide:
Specification Test Results
Related Information
Certificates
Search by Lot Number9039 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
sds_BAKR9039
SDS
spec_JT9039-10 spec_JT9039-12
Product specification
Customers Who Bought This Also Bought