Ammonium acetate ≥97% for HPLC, J.T.Baker®
Catalog #: CAJT0599-8
Supplier: VWR International
CAS Number: 631-61-8
MOC: Non-Regulated, With InformationCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Poly bottle, white, blue T/E cap
- Size:1 kg
- Supplier No.:0599-08
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (CH₃COONH₄) (by acid-base titration):≥97 %
- Insoluble Matter:≤0.005 %
- Residue after Ignition:≤0.010 %
- pH of 5% Solution at 25°C:6.7 - 7.3
- Nitrate (NO₃):≤0.001 %
- Sulfate (SO₄):≤0.001 %
- UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 350 nm:≤0.010
- UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 280 nm:≤0.010
- UV Absorbance (1 M Aqueous Solution) (1.00-cm cell vs. water) - 254 nm:≤0.020
- Chloride (Cl):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
About this item
Synonyms: Acetic acid ammonium salt
Melting Pt:114 °C
Density:1.17 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97
:Management of Change (MOC) category = NI
Flash Pt:136 °C
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 114 °C
:Flash Pt:
:Flash Pt: 136 °C
:M.W. 77.08 g/mol
Merck Index:13,00495
MW:77.08 g/mol
MDL Number:MFCD00013066
EINECS:211-162-9
:GHS07
Formula:H₃CCOONH₄
Storage Temperature:Ambient
:M.O.C. Category NI
CAS Number:631-61-8
Customers Who Bought This Also Bought