To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:HO-1
- Clonality:Polyclonal
- Gene ID:3162
- Host:Rabbit
- Isotype:IgG (H, L)
- Reactivity:Human,Rat,Mouse
- Conjugation:Unconjugated
- Flow cytometry:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µl
- Storage buffer:0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
- Molecular weight:32 kDa
- Storage temperature:Store at –20 °C for 12 months.
- Concentration:1 µg/µl
- Purification:Purified by Protein A
- Packaging:Vial
Specifications
About this item
The hemeoxygenase-1 calls that the hemoglobin oxidizes to synthesize the enzyme again-1( hemeoxygenase-1, HO-1) is the catalyst enzyme that a kind of hemoglobin declines the solution, under the NADPH and the cell dye P-450 revivification enzymes and the member oxygen functions, the catalyst HO-1 hemoglobin declines the solution as the courage green vegetable, CO and irons, the former revivification has the very strong anti- to oxidize the ability after become the red vegetable of courage , the latter is a kind of important letter to make the member.
Type: Primary
Antigen: HO-1
Clonality: Polyclonal
Conjugation: Unconjugated
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: Human, Mouse, Rat