CP100™ Nitrile Gloves, Cardinal
Supplier: Cardinal Health
Certificates
About this item
Clean-processed nitrile gloves offer excellent strength and a comfortable fit.
- Clean processed, powder free
- Excellent strength, textured grip, comfortable fit
- Ambidextrous, beaded cuff
ESD properties and 29.2 cm (11¹/₂") length make gloves ideal for many cleanroom applications. Textured surface provides a good grip. Beaded cuff to prevent rolldown. Color: white.
Ordering information: Packaged 100 gloves per double poly bag, 1000 gloves per case.
Specifications
- Catalog No:
- CAPH2Y1853
- CAPH2Y1852
- CAPH2Y1851
- CAPH2Y1849
- CAPH2Y1850
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
Specifications
Related Information
Certificates
Search by Lot NumberCardinal Health Industrial Gloves 031350W Cardinal Health™ Cleanroom Gloves Focus Safety Volume 1 Issue 1 Safety Essentials Volume 2 VWR Production insight V1
Product Literature
PH2Y1850_MY31BK3H002 PH2Y1850_MY31BK3J002 PH2Y1851_MY31BK3H003 PH2Y1852_MY31BK3H004
Conformity declaration
Customers Who Bought This Also Bought