Zinc sulfate heptahydrate, granular USP, FCC, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Formula:ZnSO₄·7H₂O
MW:287.54 g/mol
Melting Pt:100 °C
Density:1.97 g/cm³ (20 °C)
MDL Number:MFCD00149894
CAS Number:7446-20-0
ADR:9,III
Merck Index:12,10293
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 100 °C
:Flash Pt:
:Flash Pt:
:M.W. 287.54 g/mol
Storage Temperature:Ambient
EINECS:231-793-3
UN:3077
:9
:III
:GHS05
:GHS07
:GHS09
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- Food GMP Manufactured Product:
- GMP Manufactured Product:
- Meets F.C.C. Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- Assay ((as ZnSO₄)):55.6 - 61.0 %
- Assay (ZnSO₄·7H₂O):99.0 - 108.7 %
- Identification A:Passes Test
- Identification B:Passes Test
- Appearance:Passes Test
- Acidity:Passes Test
- FCC - Alkalies and Alkaline Earths:≤0.5 %
- USP - Alkalies and Alkaline Earths:≤0.9 %
- Arsenic (As):≤3 ppm
- FCC - Iron (Fe):≤0.003 %
- FCC - Lead (Pb)(FCC):≤4 mg/kg
- USP - Lead (Pb):≤0.002 %
- FCC - Cadmium (Cd):≤2 mg/kg
- FCC - Mercury (Hg):≤5 ppm
- FCC - Selenium (Se):≤0.003 %
- Solubility:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
Specification Test Results
Related Information
Certificates
Search by Lot Number8872 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_MK887207 spec_MK887220 spec_MK887224
Product specification
Customers Who Bought This Also Bought