Zinc oxide 99.0-100.5% (excluding ignited substances) USP, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Zinci oxidum
Formula:ZnO
MW:81.39 g/mol
Melting Pt:1975 °C
Density:5.61 g/cm³ (25 °C)
MDL Number:MFCD00011300
CAS Number:1314-13-2
ADR:9,III
Merck Index:13,10200
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 1975 °C
:Flash Pt:
:Flash Pt:
:M.W. 81.39 g/mol
Storage Temperature:Ambient
EINECS:215-222-5
UN:3077
REACH:01-2119463881-32
:9
:III
:GHS09
:GHS07
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Assay (ZnO) (ignited basis):98.0 - 102.0 %
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Alkalinity:Passes Test
- USP - Loss on Ignition:≤1.0 %
- USP - Carbonate and Color of Solution:Passes Test
- USP - Arsenic (As):≤6 ppm
- USP - Lead (Pb):Passes Test
- USP - Iron and Other Heavy Metals:Passes Test
- Solubility in Dilute Sulfuric Acid:Passes Test
- Lead (Pb):≤0.0010 %
- Cadmium (Cd):≤0.0015 %
- Iron (Fe):≤0.0020 %
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Metallic Residues: No metal catalysts or metal reagents, as defined by:
- EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of:
- this material.:
Specification Test Results
Related Information
Certificates
Search by Lot Number8824 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
spec_JT8824-24
Product specification
Customers Who Bought This Also Bought